Learn More
Potassium clavulanate cellulose, MedChemExpress
MedChemExpress Potassium clavulanate cellulose is a mixture of potassium clavulanate and cellulose, is a beta-lactamase inhibitor.
Brand:  Medchem Express HY-19964
Encompass_Preferred
 View more versions of this product
Product Code. 16421146
  • 287.55 € / 200mg
Please to purchase this item. Need a web account? Register with us today!
Explore more special offers
Description
Description
Potassium clavulanate cellulose is a mixture of potassium clavulanate and cellulose, is a beta-lactamase inhibitor.
Specifications
Specifications
Potassium clavulanate cellulose
Research
200 mg
DMSO : 1 mg/mL (ultrasonic and warming and heat to 80°C) ∣H2O : < 0.1 mg/mL (insoluble)
Research
COVID-19-immunoregulation
Off-White
C8H9NO5K.(C6H10O5)n
Potassium clavulanate:cellulose (1:1)
O[C@H]1[C@H](O[C@H]2[C@H](O)[C@@H](O)[C@H](C)[C@@H](CO)O2)[C@@H](CO)O[C@@H](OC)[C@@H]1O.O=C([C@@H](/C(O[C@]3([H])C4)=C/CO)N3C4=O)O[K].[n]
Solid