missing translation for 'onlineSavingsMsg'
Learn More

Potassium clavulanate cellulose, MedChemExpress

MedChemExpress Potassium clavulanate cellulose is a mixture of potassium clavulanate and cellulose, is a beta-lactamase inhibitor.

79.65 € - 637.20 €

Specifications

Molecular Formula C8H9NO5K.(C6H10O5)n
View More Specs

Products 5
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16477766
Encompass_Preferred
View Documents
Medchem Express
HY-19964
100 mg
175.50 €
100mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16423917
Encompass_Preferred
View Documents
Medchem Express
HY-19964
50 mg
112.05 €
50mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16421146
Encompass_Preferred
View Documents
Medchem Express
HY-19964
200 mg
287.55 €
200mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16457988
Encompass_Preferred
View Documents
Medchem Express
HY-19964
10 mg
79.65 €
10mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16441906
Encompass_Preferred
View Documents
Medchem Express
HY-19964
500 mg
637.20 €
500mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Potassium clavulanate cellulose is a mixture of potassium clavulanate and cellulose, is a beta-lactamase inhibitor.
Specifications

Specifications

C8H9NO5K.(C6H10O5)n
Videos
Safety and Handling

Safety and Handling

missing translation for 'shelfLife' : 4°C, sealed storage, away from moisture∣In solvent : -80°C, 6 months∣-20°C, 1 month (sealed storage, away from moisture)

Recommended Storage : 4°C, sealed storage, away from moisture∣In solvent : -80°C, 6 months∣-20°C, 1 month (sealed storage, away from moisture)

SDS
Documents

Documents

Product Certifications

Research purposes only