Learn More
Potassium clavulanate cellulose, MedChemExpress
MedChemExpress Potassium clavulanate cellulose is a mixture of potassium clavulanate and cellulose, is a beta-lactamase inhibitor.
Brand: Medchem Express HY-19964
This item is not returnable.
View return policy
Description
Potassium clavulanate cellulose is a mixture of potassium clavulanate and cellulose, is a beta-lactamase inhibitor.Specifications
Potassium clavulanate cellulose | |
Research | |
10 mg | |
DMSO : 1 mg/mL (ultrasonic and warming and heat to 80°C) ∣H2O : < 0.1 mg/mL (insoluble) | |
Research | |
COVID-19-immunoregulation |
Off-White | |
C8H9NO5K.(C6H10O5)n | |
Potassium clavulanate:cellulose (1:1) | |
O[C@H]1[C@H](O[C@H]2[C@H](O)[C@@H](O)[C@H](C)[C@@H](CO)O2)[C@@H](CO)O[C@@H](OC)[C@@H]1O.O=C([C@@H](/C(O[C@]3([H])C4)=C/CO)N3C4=O)O[K].[n] | |
Solid |
Research purposes only