missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Q-VD-OPH Inhibitor Peptide
Applications: Functional, In vitro assay
Brand: Novus Biologicals™ NBP2-29391-3MG
This item is not returnable.
View return policy
Description

Specifications
Human | |
Functional, In vitro assay | |
3 mg | |
513 | |
Semi-solid |
Off-white | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
Q-VD-OPH Inhibitor Peptide | |
Q-VD-OPH |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Novus Biologicals™ Q-VD-OPH Inhibitor Peptide > 3mg
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction