Learn More
Lenz Laborglasintrumente Reaction Vessels, Spherical, with Thermostatic Jacket
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
Brand: Lenz Laborglasintrumente 06021158
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
- Without graduation
- With laboratory flange LF
- With groove, with two connectors (with threads GL 14 /GL 18 or ball flanges KF 15)
- Made of DURAN™ tubing
- Other sizes on request
Spezifikation
0.5 L | |
180 mm | |
1 Ea. |
Duran Glass | |
Lab Flange 60 LF, Connector KF 15 | |
131 mm |