Learn More
Lenz Laborglasintrumente Reaction Vessels, Spherical, with Thermostatic Jacket
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
1266.70 € - 6900.60 €
Product Code | Brand | Height (Metric) | Includes | Diameter (Metric) Outer | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Height (Metric) | Includes | Diameter (Metric) Outer | Price | Quantity & Availability | |||
10710435
|
Lenz Laborglasintrumente
06020273 |
260 mm | Lab Flange 100 LF, Connector GL 18 | 206 mm |
1266.70 €
Each |
Estimated Shipment: 03-12-2024
Log in to see stock. |
|||
11502274
|
Lenz Laborglasintrumente
06020285 |
340 mm | Lab Flange 100 LF, Connector GL 18 | 280 mm |
1929.20 €
Each |
Estimated Shipment: 03-12-2024
Log in to see stock. |
|||
11502284
|
Lenz Laborglasintrumente
06021585 |
340 mm | Lab Flange 200 LF, Connector KF 15 | 280 mm |
4160.50 €
Each |
Estimated Shipment: 03-12-2024
Log in to see stock. |
|||
11512274
|
Lenz Laborglasintrumente
06020485 |
355 mm | Lab Flange 150 LF, Connector GL 18 | 280 mm |
2385.00 €
Each |
Estimated Shipment: 03-12-2024
Log in to see stock. |
|||
11512284
|
Lenz Laborglasintrumente
06021588 |
420 mm | Lab Flange 200 LF, Connector KF 15 | 350 mm |
6900.60 €
Each |
Estimated Shipment: 03-12-2024
Log in to see stock. |
|||
11542274
|
Lenz Laborglasintrumente
06021149 |
155 mm | Lab Flange 60 LF, Connector KF 15 | 105 mm |
1595.30 €
Each |
Estimated Shipment: 03-12-2024
Log in to see stock. |
|||
11552274
|
Lenz Laborglasintrumente
06021158 |
180 mm | Lab Flange 60 LF, Connector KF 15 | 131 mm |
1627.10 €
Each |
Estimated Shipment: 03-12-2024
Log in to see stock. |
|||
11562274
|
Lenz Laborglasintrumente
06021170 |
215 mm | Lab Flange 60 LF, Connector KF 15 | 165 mm |
1749.00 €
Each |
Estimated Shipment: 03-12-2024
Log in to see stock. |
|||
11572264
|
Lenz Laborglasintrumente
06020158 |
180 mm | Lab Flange 60 LF, Connector GL 14 | 131 mm |
1441.60 €
Each |
Estimated Shipment: 03-12-2024
Log in to see stock. |
|||
11572274
|
Lenz Laborglasintrumente
06021273 |
260 mm | Lab Flange 100 LF, Connector KF 15 | 206 mm |
1690.70 €
Each |
Estimated Shipment: 03-12-2024
Log in to see stock. |
|||
11582264
|
Lenz Laborglasintrumente
06020170 |
215 mm | Lab Flange 60 LF, Connector GL 14 | 165 mm |
1505.20 €
Each |
Estimated Shipment: 03-12-2024
Log in to see stock. |
|||
11582274
|
Lenz Laborglasintrumente
06021279 |
295 mm | Lab Flange 100 LF, Connector KF 15 | 236 mm |
1945.10 €
Each |
Estimated Shipment: 03-12-2024
Log in to see stock. |
|||
11592264
|
Lenz Laborglasintrumente
06020279 |
295 mm | Lab Flange 100 LF, Connector GL 18 | 236 mm |
1727.80 €
Each |
Estimated Shipment: 03-12-2024
Log in to see stock. |
|||
11592274
|
Lenz Laborglasintrumente
06021285 |
340 mm | Lab Flange 100 LF, Connector KF 15 | 280 mm |
2173.00 €
Each |
Estimated Shipment: 03-12-2024
Log in to see stock. |
|||
11913701
|
Lenz Laborglasintrumente
06020149 |
155 mm | Lab Flange 60 LF, Connector GL 14 | 105 mm |
1409.80 €
Each |
Estimated Shipment: 03-12-2024
Log in to see stock. |
|||
Description
- Without graduation
- With laboratory flange LF
- With groove, with two connectors (with threads GL 14 /GL 18 or ball flanges KF 15)
- Made of DURAN™ tubing
- Other sizes on request
Specifications
Duran Glass |