Learn More
Thermo Scientific™ Orion™ 2230XP Silica Analyzer Reagents and Standards
The reagents and calibration standards you need to operate and maintain the Thermo Scientific™ Orion™ 2230XP Silica Analyzer.
Brand:  Thermo Scientific™ 2230RE
 View more versions of this product
Product Code. 15914267
  • 499.00 € / 1 set
Please to purchase this item. Need a web account? Register with us today!
Explore more special offers
This item is not returnable. View return policy
Description
Description
Ensure you have the reagents and calibration standards you need to operate and maintain the Thermo Scientific™ Orion™ 2230XP Silica Analyzer. Reagents kit contains all three reagents required for operation, and calibration standards available in individual bottles. For use with the Orion 2230XP Silica Analyzer.
Warranty: Out of box
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Reagents 1,2 and 3
5°C to 45°C
Reagents and Standard
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
Signal Word
  • Warning
Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P202-Do not handle until all safety precautions have been read and understood.
  • P321-Specific treatment (see on this label).
Supplemental information
  • MIXTURE LIST-Contains : sodium metasilicatenonahydrate
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Thermo Scientific™ Orion™ 2230XP Silica Analyzer Reagents and Standards >
Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.