missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Bone sialoprotein 1

A DNA sequence encoding the Homo sapiens (Human) Bone sialoprotein 2, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6187-ec-1mg

 View more versions of this product

Product Code. 16071420

  • 2485.00 € / 1mg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Specifications

Specifications

NP_004958.2
Bone sialoprotein 1
Research Use Only
IBSP
Recombinant Protein
E. coli
AIQLPKKAGD ITNKATKEKE SDEEEEEEEE GNENEESEAE VDENEQGING TSTNSTEAEN GNGSSGGDNG EEGEEESVTG ANAEDTTETG RQGKGTSKTT TSPNGGFEPT TPPQVYRTTS PPFGKTTTVE YEGEYEYTGA NEYDNGYEIY ESE
Greater than 80% as determined by SDS-PAGE.
3381
1 mg
Not determined.
Partial (see sequence information for more details).
Human
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions