missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Collagen alpha-1(XVII) chain Protein
A DNA sequence encoding the Homo sapiens (Human) Collagen alpha-1(XVII) chain, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8920-ec-500ug
This item is not returnable.
View return policy
Specifications
NP_000485.3 | |
Collagen alpha-1(XVII) chain Protein | |
Research Use Only | |
COL17A1 | |
Recombinant Protein | |
E. coli | |
YLTSPDVRSF IVGPPGPPGP QGPPGDSRLL STDASHSRGS SSSSHSSSVR RGSSYSSSMS TGGGGAGSLG AGGAFGEAAG DRGPYGTDIG PGGGYGAAAE GGMYAGNGGL LGADFAGDLD YNELAVRVSE SMQRQGLLQG MAYTVQGPPG QPGPQGPPGI SKVFSAYSNV TADLMDFFQT YGAIQGPPGQ KGEMGTPGPK GDRGPAGPPG HPGPPGPRGH KGEKGDKGDQ VYAGRRRRRS IAVKP | |
Greater than 90% as determined by SDS-PAGE. |
1308 | |
500 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |