missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Collagen alpha-1(XVII) chain Protein

A DNA sequence encoding the Homo sapiens (Human) Collagen alpha-1(XVII) chain, was expressed in the hosts and tags indicated.

160.00 € - 1855.00 €

Specifications

Accession Number NP_000485.3
Gene ID (Entrez) 1308
Name Collagen alpha-1(XVII) chain Protein
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
View More Specs

Products 6
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16007181
View Documents Promotion Details
enQuireBio™
QP8920-ec-10ug
10 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16047181
View Documents Promotion Details
enQuireBio™
QP8920-ec-50ug
50 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16097171
View Documents Promotion Details
enQuireBio™
QP8920-ec-100ug
100 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16027181
View Documents Promotion Details
enQuireBio™
QP8920-ec-200ug
200 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16037181
View Documents Promotion Details
enQuireBio™
QP8920-ec-500ug
500 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16017181
View Documents Promotion Details
enQuireBio™
QP8920-ec-1mg
1 mg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

Specifications

Specifications

NP_000485.3
Collagen alpha-1(XVII) chain Protein
Not determined.
Partial (see sequence information for more details).
Human
His
Tris-based buffer, 50% glycerol
1308
Research Use Only
COL17A1
Recombinant Protein
E. coli
YLTSPDVRSF IVGPPGPPGP QGPPGDSRLL STDASHSRGS SSSSHSSSVR RGSSYSSSMS TGGGGAGSLG AGGAFGEAAG DRGPYGTDIG PGGGYGAAAE GGMYAGNGGL LGADFAGDLD YNELAVRVSE SMQRQGLLQG MAYTVQGPPG QPGPQGPPGI SKVFSAYSNV TADLMDFFQT YGAIQGPPGQ KGEMGTPGPK GDRGPAGPPG HPGPPGPRGH KGEKGDKGDQ VYAGRRRRRS IAVKP
Greater than 90% as determined by SDS-PAGE.
Videos
SDS
Documents

Documents

Product Certifications