Learn More
R&D Systems™ Recombinant Human IL-23 Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
Brand:  R&D Systems™ 1290-IL-010/CF
388.76 EUR valid until 2024-12-16
Use promo code "21630" to get your promotional price.
 View more versions of this product
Product Code. 18118640
  • 516.00 € / 10µg
Please to purchase this item. Need a web account? Register with us today!
Explore more special offers
Description
Description
The Recombinant Human IL-23 Protein is derived from Sf 21 (baculovirus). The Recombinant Human IL-23 Protein has been validated for the following applications: Bioactivity.
Specifications
Specifications
Supplied as a 0.2μm filtered solution in MES, NaCl and EDTA.
55.0 kDa (IL-23 single chain), 36.5 kDa (p40), 18.5 kDa (p19)
10 μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after opening. 3 months, -20 to -70° C under sterile conditions after opening.
Recombinant
51561
IL-23
Spodoptera frugiperda,Sf 21 (baculovirus)-derived human IL-23 protein, (N-terminus) Human IL-23 p40 (Ile23-Ser328) Accession &Num; P29460 GSGSSRGGSGSGGSGGGGSKL Human IL-23 p19 (Arg20-Pro189) Accession &Num; AAG37232 (C-terminus)
Unconjugated
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.