missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human IL-23 Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
516.00 €
Specifications
Gene ID (Entrez) | 51561 |
---|---|
Name | IL-23 |
Quantity | 10 μg |
Source | Spodoptera frugiperda,Sf 21 (baculovirus)-derived human IL-23 protein, (N-terminus) Human IL-23 p40 (Ile23-Ser328) Accession &Num; P29460 GSGSSRGGSGSGGSGGGGSKL Human IL-23 p19 (Arg20-Pro189) Accession &Num; AAG37232 (C-terminus) |
Conjugate | Unconjugated |
Product Code | Brand | Formulation | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Formulation | Price | Quantity & Availability | |||||
18117540
|
R&D Systems™
1290-IL-010 |
Lyophilized from a 0.2μm filtered solution in MES, NaCl and EDTA with BSA as a carrier protein. |
516.00 €
10µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18118640
|
R&D Systems™
1290-IL-010/CF |
Supplied as a 0.2μm filtered solution in MES, NaCl and EDTA. |
516.00 €
10µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The Recombinant Human IL-23 Protein is derived from Sf 21 (baculovirus). The Recombinant Human IL-23 Protein has been validated for the following applications: Bioactivity.Specifications
51561 | |
10 μg | |
Unconjugated | |
95%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
IL-23 | |
Spodoptera frugiperda,Sf 21 (baculovirus)-derived human IL-23 protein, (N-terminus) Human IL-23 p40 (Ile23-Ser328) Accession &Num; P29460 GSGSSRGGSGSGGSGGGGSKL Human IL-23 p19 (Arg20-Pro189) Accession &Num; AAG37232 (C-terminus) | |
Recombinant |