missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human MAD2L1BP Protein

A cDNA sequence encoding the MAD2L1BP was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP12617-1mg

 View more versions of this product

Product Code. 15941009

  • 8190.00 € / 1mg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Specifications

Specifications

MAD2L1BP Protein
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSHMAAPEA EVLSSAAVPD LEWYEKSEET HASQIELLET SSTQEPLNAS EAFCPRDCMV PVVFPGPVSQ EGCCQFTCEL LKHIMYQRQQ LPLPYEQLKH FYRKPSPQAE EMLKKKPRAT TEVSSRKCQQ ALAELESVLS HLEDFFARTL VPRVLILLGGNALSPKEFYE LDLSLLAPYS VDQSLSTAAC LRRLFRAIFM ADAFSELQAP PLMGTVVMAQ GHRNCGEDWF RPKLNYRVPS RGHKLTVTLS CGRPSIRTTA WEDYIWFQAP VTFKGFRE
Greater than 90.0% as determined by SDS-PAGE.
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
MAD2L1BP protein solution (0.5 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 1mM DTT, 10% glycerol and 100mM NaCl.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.