missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human MAD2L1BP Protein
A cDNA sequence encoding the MAD2L1BP was constructed and used to recombinantly synthesize the protein.
137.00 € - 7875.00 €
Specifications
Name | MAD2L1BP Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15951009
|
enQuireBio™
QP12617-2ug |
2 μg |
137.00 €
2µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15931009
|
enQuireBio™
QP12617-10ug |
10 μg |
212.00 €
10µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15941009
|
enQuireBio™
QP12617-1mg |
1 mg |
7875.00 €
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
MAD2L1BP Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
MAD2L1BP protein solution (0.5 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 1mM DTT, 10% glycerol and 100mM NaCl. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSHMAAPEA EVLSSAAVPD LEWYEKSEET HASQIELLET SSTQEPLNAS EAFCPRDCMV PVVFPGPVSQ EGCCQFTCEL LKHIMYQRQQ LPLPYEQLKH FYRKPSPQAE EMLKKKPRAT TEVSSRKCQQ ALAELESVLS HLEDFFARTL VPRVLILLGGNALSPKEFYE LDLSLLAPYS VDQSLSTAAC LRRLFRAIFM ADAFSELQAP PLMGTVVMAQ GHRNCGEDWF RPKLNYRVPS RGHKLTVTLS CGRPSIRTTA WEDYIWFQAP VTFKGFRE | |
Greater than 90.0% as determined by SDS-PAGE. |