missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human MAD2L1BP Protein
A cDNA sequence encoding the MAD2L1BP was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12617-2ug
This item is not returnable.
View return policy
Specifications
MAD2L1BP Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSHMAAPEA EVLSSAAVPD LEWYEKSEET HASQIELLET SSTQEPLNAS EAFCPRDCMV PVVFPGPVSQ EGCCQFTCEL LKHIMYQRQQ LPLPYEQLKH FYRKPSPQAE EMLKKKPRAT TEVSSRKCQQ ALAELESVLS HLEDFFARTL VPRVLILLGGNALSPKEFYE LDLSLLAPYS VDQSLSTAAC LRRLFRAIFM ADAFSELQAP PLMGTVVMAQ GHRNCGEDWF RPKLNYRVPS RGHKLTVTLS CGRPSIRTTA WEDYIWFQAP VTFKGFRE | |
Greater than 90.0% as determined by SDS-PAGE. |
2 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
MAD2L1BP protein solution (0.5 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 1mM DTT, 10% glycerol and 100mM NaCl. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction