missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Protein cereblon Protein
A DNA sequence encoding the Homo sapiens(Human) Protein cereblon, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP10011-iv-50ug
This item is not returnable.
View return policy
Specifications
NP_001166953.1 | |
Protein cereblon Protein | |
Research Use Only | |
Crbn | |
Recombinant Protein | |
E. coli | |
MAGEGDQQDA AHNMGNHLPL LPAESEEEDE MEVEDQDSKE AKKPNIINFD TSLPTSHTYL GADMEEFHGR TLHDDDSCQV IPVLPQVMMI LIPGQTLPLQ LFHPQEVSMV RNLIQKDRTF AVLAYSNVQE REAQFGTTAE IYAYREEQDF GIEIVKVKAI GRQRFKVLEL RTQSDGIQQA KVQILPECVL PSTMSAVQLE SLNKCQIFPS KPVSREDQCS YKWWQKYQKR KFHCANLTSW PRWLYSLYDA ETLMDRIKKQ LREWDENLKD DSLPSNPIDF SYRVAACLPI DDVLRIQLLK IGSAIQRLRC ELDIMNKCTS LCCKQCQETE ITTKNEIFSL SLCGPMAAYV NPHGYVHETL TVYKACNLNL IGRPSTEHSW FPGYAWTVAQ CKICASHIGW KFTATKKDMS PQKFWGLTRS ALLPTIPDTE DEISPDKVIL CL | |
Greater than 80% as determined by SDS-PAGE. |
51185 | |
50 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |