missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Protein cereblon Protein
A DNA sequence encoding the Homo sapiens(Human) Protein cereblon, was expressed in the hosts and tags indicated.
469.00 € - 1705.00 €
Specifications
Accession Number | NP_001166953.1 |
---|---|
Gene ID (Entrez) | 51185 |
Name | Protein cereblon Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15962748
|
enQuireBio™
QP10011-iv-10ug |
10 μg | N/A | ||||||
15972748
|
enQuireBio™
QP10011-iv-50ug |
50 μg | N/A | ||||||
15952748
|
enQuireBio™
QP10011-iv-100ug |
100 μg |
1705.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
NP_001166953.1 | |
Protein cereblon Protein | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
51185 | |
Research Use Only | |
Crbn | |
Recombinant Protein | |
E. coli | |
MAGEGDQQDA AHNMGNHLPL LPAESEEEDE MEVEDQDSKE AKKPNIINFD TSLPTSHTYL GADMEEFHGR TLHDDDSCQV IPVLPQVMMI LIPGQTLPLQ LFHPQEVSMV RNLIQKDRTF AVLAYSNVQE REAQFGTTAE IYAYREEQDF GIEIVKVKAI GRQRFKVLEL RTQSDGIQQA KVQILPECVL PSTMSAVQLE SLNKCQIFPS KPVSREDQCS YKWWQKYQKR KFHCANLTSW PRWLYSLYDA ETLMDRIKKQ LREWDENLKD DSLPSNPIDF SYRVAACLPI DDVLRIQLLK IGSAIQRLRC ELDIMNKCTS LCCKQCQETE ITTKNEIFSL SLCGPMAAYV NPHGYVHETL TVYKACNLNL IGRPSTEHSW FPGYAWTVAQ CKICASHIGW KFTATKKDMS PQKFWGLTRS ALLPTIPDTE DEISPDKVIL CL | |
Greater than 80% as determined by SDS-PAGE. |