missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human SF20 Protein

A cDNA sequence encoding the SF20 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP13474-1mg

 View more versions of this product

Product Code. 15963689

  • 2725.00 € / 1mg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Specifications

Specifications

SF20 Protein
Research Use Only
Recombinant Protein
E. coli
MAAPSGGWNG VGASLWAALL LGAVALRPAE AVSEPTTVAF DVRPGGVVHS FSHNVGPGDK YTCMFTYASQ GGTNEQWQMS LGTSEDHQHF TCTIWRPQGK SYLYFTQFKA EVRGAEIEYA MAYSKAAFER ESDVPLKTEE FEVTKTAVAH RPGAFKAELS KLVIVAKASR TEL
Greater than 95.0% by SDS-PAGE.
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
C9orf10 is supplied in 20mM Tris HCL pH 8 and 20% glycerol.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions