missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human SF20 Protein
A cDNA sequence encoding the SF20 was constructed and used to recombinantly synthesize the protein.
137.00 € - 2725.00 €
Specifications
Name | SF20 Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15953689
|
enQuireBio™
QP13474-10ug |
10 μg |
137.00 €
10µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15973689
|
enQuireBio™
QP13474-50ug |
50 μg |
212.00 €
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15963689
|
enQuireBio™
QP13474-1mg |
1 mg |
2725.00 €
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
SF20 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
C9orf10 is supplied in 20mM Tris HCL pH 8 and 20% glycerol. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MAAPSGGWNG VGASLWAALL LGAVALRPAE AVSEPTTVAF DVRPGGVVHS FSHNVGPGDK YTCMFTYASQ GGTNEQWQMS LGTSEDHQHF TCTIWRPQGK SYLYFTQFKA EVRGAEIEYA MAYSKAAFER ESDVPLKTEE FEVTKTAVAH RPGAFKAELS KLVIVAKASR TEL | |
Greater than 95.0% by SDS-PAGE. |