missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human SNRPD1 Protein
A cDNA sequence encoding the SNRPD1 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP13547-1mg
This item is not returnable.
View return policy
Specifications
SNRPD1 Protein | |
Research Use Only | |
Recombinant Protein | |
Baculovirus | |
MKLVRFLMKL SHETVTIELK NGTQVHGTIT GVDVSMNTHL KAVKMTLKNR EPVQLETLSI RGNNIRYFIL PDSLPLDTLL VDVEPKVKSK KREAVAGRGR GRGRGRGRGR GRGRGGPRR | |
Greater than 80% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
SNRPD1 is supplied in 20mM HEPES buffer pH 7.6, 250mM NaCl and 20% glycerol. |