missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human SNRPD1 Protein
A cDNA sequence encoding the SNRPD1 was constructed and used to recombinantly synthesize the protein.
137.00 € - 7875.00 €
Specifications
Name | SNRPD1 Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15973929
|
enQuireBio™
QP13547-2ug |
2 μg |
137.00 €
2µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15953929
|
enQuireBio™
QP13547-10ug |
10 μg |
212.00 €
10µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15963929
|
enQuireBio™
QP13547-1mg |
1 mg |
7875.00 €
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
SNRPD1 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
SNRPD1 is supplied in 20mM HEPES buffer pH 7.6, 250mM NaCl and 20% glycerol. |
Research Use Only | |
Recombinant Protein | |
Baculovirus | |
MKLVRFLMKL SHETVTIELK NGTQVHGTIT GVDVSMNTHL KAVKMTLKNR EPVQLETLSI RGNNIRYFIL PDSLPLDTLL VDVEPKVKSK KREAVAGRGR GRGRGRGRGR GRGRGGPRR | |
Greater than 80% as determined by SDS-PAGE. |