missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human SNRPD1 Protein

A cDNA sequence encoding the SNRPD1 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP13547-2ug

 View more versions of this product

Product Code. 15973929

  • 137.00 € / 2µg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Specifications

Specifications

SNRPD1 Protein
Research Use Only
Recombinant Protein
Baculovirus
MKLVRFLMKL SHETVTIELK NGTQVHGTIT GVDVSMNTHL KAVKMTLKNR EPVQLETLSI RGNNIRYFIL PDSLPLDTLL VDVEPKVKSK KREAVAGRGR GRGRGRGRGR GRGRGGPRR
Greater than 80% as determined by SDS-PAGE.
2 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
SNRPD1 is supplied in 20mM HEPES buffer pH 7.6, 250mM NaCl and 20% glycerol.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions