missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Rat RAP Protein

A cDNA sequence encoding the RAP was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP13264-1mg

 View more versions of this product

Product Code. 15943019

  • 4410.00 € / 1mg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Specifications

Specifications

RAP Protein
Research Use Only
Recombinant Protein
E. coli
MAPLRDRVSTLPRLQLLVLLLLPLLLVPQPIAGHGGKYSREKNEPEMAAKRESGEEFRME KLNQLWEKAKRLHLSPVRLAELHSDLKIQERDELNWKKLKVEGLDGDGEKEAKLVHNLNV ILARYGLDGRKDTQTVHSNALNEDTQDELGDPRLEKLWHKAKTSGKFSSEELDKLWREFL HYKEKIHEYNVLLDTLSRAEEGYENLLSPSDMTHIKSDTLASKHSELKDRLRSINQGLDR LRKVSHQGYGPATEFEEPRVIDLWDLAQSANFTEKELESFREELKHFEAKIEKHNHYQKQ LEISHQKLKHVESIGDPEHISRNKEKYVLLEEKTKELGYKVKKHLQDLSSRVSRARHNEL
Greater than 95.0% as determined by SDS-PAGE.
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Rat
His
The protein (1 mg/ml) was lyophilized after from a sterile solution containing TBS pH 7.5, 0.1% BSA and 0.09% NaN3.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions