missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat RAP Protein
A cDNA sequence encoding the RAP was constructed and used to recombinantly synthesize the protein.
137.00 € - 4240.00 €
Specifications
Name | RAP Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Rat |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15963019
|
enQuireBio™
QP13264-5ug |
5 μg |
137.00 €
5µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15953019
|
enQuireBio™
QP13264-20ug |
20 μg |
212.00 €
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15943019
|
enQuireBio™
QP13264-1mg |
1 mg |
4240.00 €
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
RAP Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Rat | |
His | |
The protein (1 mg/ml) was lyophilized after from a sterile solution containing TBS pH 7.5, 0.1% BSA and 0.09% NaN3. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MAPLRDRVSTLPRLQLLVLLLLPLLLVPQPIAGHGGKYSREKNEPEMAAKRESGEEFRME KLNQLWEKAKRLHLSPVRLAELHSDLKIQERDELNWKKLKVEGLDGDGEKEAKLVHNLNV ILARYGLDGRKDTQTVHSNALNEDTQDELGDPRLEKLWHKAKTSGKFSSEELDKLWREFL HYKEKIHEYNVLLDTLSRAEEGYENLLSPSDMTHIKSDTLASKHSELKDRLRSINQGLDR LRKVSHQGYGPATEFEEPRVIDLWDLAQSANFTEKELESFREELKHFEAKIEKHNHYQKQ LEISHQKLKHVESIGDPEHISRNKEKYVLLEEKTKELGYKVKKHLQDLSSRVSRARHNEL | |
Greater than 95.0% as determined by SDS-PAGE. |