missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Recombinant Rat VEGF120 Protein
Recombinant Protein
Brand: Novus Biologicals™ NBP2-76336-1000ug
This item is not returnable.
View return policy
Description
VEGF120 protein is fully biologically active when compared to standard. Measured in a cell proliferation assay using HUVEC human umbilical vein endothelial cells. The ED50 for this effect is 2-10 ng/mL.Specifications
Bioactivity | |
7422 | |
1000 μg | |
Less than 0.1 EU/μg of VEGF120 as determined by LAL method. | |
Recommended to centrifuge prior to opening. Reconstitute in sterile distilled water or aqueous buffer containing 0.1% BSA to a concentration of 0.1-1.0 mg/mL Apportion stock solutions into working aliquots and store (at <-20°C.) | |
>95%, by SDS-PAGE |
Lyophilized from a 0.2 μm filtered concentrated solution in PBS, pH 7.4. | |
MolecularWeight-theroretical: 18.5 kDa | |
Store at -20°C to -70°C as supplied. After reconstitution, store at 2 to 8 C for 1 month and (at -20°C to -70°C for long term storage. Avoid repeated freeze/thaw cycles.) | |
VEGFA | |
As a result of glycosylation, it migrates to at least two bands with molecular weights ranging from 18.5 kDa in SDS-PAGE under reducing conditions. | |
VEGF120 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction