missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Recombinant Rat VEGF120 Protein
Recombinant Protein
238.00 € - 2950.00 €
Specifications
For Use With (Application) | Bioactivity |
---|---|
Formulation | Lyophilized from a 0.2 μm filtered concentrated solution in PBS, pH 7.4. |
Gene ID (Entrez) | 7422 |
Molecular Weight (g/mol) | MolecularWeight-theroretical: 18.5 kDa |
Storage Requirements | Store at -20°C to -70°C as supplied. After reconstitution, store at 2 to 8 C for 1 month and (at -20°C to -70°C for long term storage. Avoid repeated freeze/thaw cycles.) |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18666420
|
Novus Biologicals™
NBP2-76336-500ug |
500 μg |
1805.00 €
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18646141
|
Novus Biologicals™
NBP2-76336-10ug |
10 μg |
238.00 €
10µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18684560
|
Novus Biologicals™
NBP2-76336-100ug |
100 μg |
978.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18618061
|
Novus Biologicals™
NBP2-76336-1000ug |
1000 μg |
2950.00 €
1000µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
VEGF120 protein is fully biologically active when compared to standard. Measured in a cell proliferation assay using HUVEC human umbilical vein endothelial cells. The ED50 for this effect is 2-10 ng/mL.Specifications
Bioactivity | |
7422 | |
Store at -20°C to -70°C as supplied. After reconstitution, store at 2 to 8 C for 1 month and (at -20°C to -70°C for long term storage. Avoid repeated freeze/thaw cycles.) | |
VEGFA | |
As a result of glycosylation, it migrates to at least two bands with molecular weights ranging from 18.5 kDa in SDS-PAGE under reducing conditions. | |
VEGF120 |
Lyophilized from a 0.2 μm filtered concentrated solution in PBS, pH 7.4. | |
MolecularWeight-theroretical: 18.5 kDa | |
Less than 0.1 EU/μg of VEGF120 as determined by LAL method. | |
Recommended to centrifuge prior to opening. Reconstitute in sterile distilled water or aqueous buffer containing 0.1% BSA to a concentration of 0.1-1.0 mg/mL Apportion stock solutions into working aliquots and store (at <-20°C.) | |
>95%, by SDS-PAGE |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title