Learn More
Thermo Scientific™ Consumable Parts for Integrated Hydride Kit for iCAP™ ICP-OES Systems
Ensure peak performance of your ICP-OES analyzer with dedicated consumables for analysis of hydride-generating elements.
330.65 €
Products 1
Product Code Brand dimType Description Price Quantity & Availability  
Product Code Brand dimType Description Price Quantity & Availability  
17691795
View Documents
Thermo Scientific™
842318050151
330.65 €
Each
Estimated Shipment: 28-02-2025
to see stock.
Add to basket
Description
Description
Ensure peak performance of your Thermo Scientific™ iCAP™ Series ICP-OES system using dedicated Thermo Scientific™ Consumable Parts for the Integrated Hydride Kit for ICP-OES systems.
Peak Performance with Dedicated Consumables
The replacement pump tubing and a consumables kit ensures the peak performance of hydride analysis on your iCAP Series ICP-OES System.
Consumables kit contains:
  • Pump tubing (for both, mini (95 mm) and standard peristaltic pump (152 mm))
  • Teflon membrane
  • O-ring
  • Replacement tubing
  • Sample probe