missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Roll of Not Drinking Water Warning Labels

Brand:  Brady™ 227380

 View more versions of this product

Product Code. 10535704

  • 372.00 € / 1 roll
Estimated Shipment: 11-04-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Specifications

Specifications

PIC 204 RL-DIA 100-B7541
Laminated Polyester
Permanent Acrylic
0.099 mm
Round
Black on White
Gloss
250/Pk.
100 mm
-40 °C-110 °C
Global
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Brady™ Roll of Not Drinking Water Warning Labels > Dia.: 100mm

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.