Learn More
Poulten & Graf GmbH FORTUNA™ OPTIMA™ Glass and Metal Syringe with Record Tip
Poulten & Graf GmbH FORTUNA™ OPTIMA™ Glass and Metal Syringe with Record Tip is used to measure and dispense fluids.
Brand: Poulten & Graf GmbH 7 20041
This item is not returnable.
View return policy
Description
- With recorder tip
- With steel piston
- Interchangable
- For technical use only
Specifications
Autoclavable | |
Clear | |
Yes | |
Reusable | |
Glass and Metal Syringe | |
20 mL | |
Yes |
Neutral Glass, Neutral Glass (Barrel), Nickel-plated Brass (Tip), Steel (Plunger) | |
Glass and Metal Syringe with Record tip | |
Glass and Metal Syringe | |
Amber | |
1 | |
Record Tip |