Learn More
Keystone Cleanroom Products Self-Sealing 1421B Tyvek™ Autoclave Bag with Steam Indicator, 0 to 12 in. Wide
The Self-Sealing 1421B Tyvek™ Autoclave Bag with Steam Indicator is designed for the steam sterilization of pharmaceutical equipment change parts. These bags are closed via the convenient and robust self-sealing closure system.
1469.44 € - 3470.77 €
Specifications
Product Type Autoclave Bag
Autoclavable Autoclavable
Certifications/Compliance Certificate of Conformance (COC)
Closure Type Self-seal
Color White
View More Specs
Products 44
Product Code Brand Dimensions (L x W) Price Quantity & Availability  
Product Code Brand Dimensions (L x W) Price Quantity & Availability  
30020166
View Documents Promotion Details
Keystone Cleanroom Products
6STK0404SS
4 x 4 in.
1717.35 €
Pack of 250
Please to purchase this item. Need a web account? Register with us today!
30020333
View Documents Promotion Details
Keystone Cleanroom Products
6STK0707SS
7 x 7 in.
1915.69 €
Pack of 250
Please to purchase this item. Need a web account? Register with us today!
30020175
View Documents Promotion Details
Keystone Cleanroom Products
6STK0608SS
8 x 6 in.
1469.44 €
Pack of 250
Please to purchase this item. Need a web account? Register with us today!
30020123
View Documents Promotion Details
Keystone Cleanroom Products
6STK0409SS
9 x 4 in.
3209.33 €
Pack of 500
Please to purchase this item. Need a web account? Register with us today!
30020198
View Documents Promotion Details
Keystone Cleanroom Products
6STK0709SS
9 x 7 in.
1951.75 €
Pack of 250
Please to purchase this item. Need a web account? Register with us today!
30020372
View Documents Promotion Details
Keystone Cleanroom Products
6STK09-512SS
9.5 x 12 in.
2028.37 €
Pack of 250
Please to purchase this item. Need a web account? Register with us today!
30020272
View Documents Promotion Details
Keystone Cleanroom Products
6STK0610SS
10 x 6 in.
3407.67 €
Pack of 500
Please to purchase this item. Need a web account? Register with us today!
30020119
View Documents Promotion Details
Keystone Cleanroom Products
6STK0811SS
11 x 8 in.
2159.10 €
Pack of 250
Please to purchase this item. Need a web account? Register with us today!
30020056
View Documents Promotion Details
Keystone Cleanroom Products
6STK0612SS
12 x 6 in.
3470.77 €
Pack of 500
Please to purchase this item. Need a web account? Register with us today!
30020111
View Documents Promotion Details
Keystone Cleanroom Products
6STK1212SS
12 x 12 in.
2249.23 €
Pack of 250
Please to purchase this item. Need a web account? Register with us today!
30020264
View Documents Promotion Details
Keystone Cleanroom Products
6STK0713SS
13 x 7 in.
2010.35 €
Pack of 250
Please to purchase this item. Need a web account? Register with us today!
30020134
View Documents Promotion Details
Keystone Cleanroom Products
6STK1214SS
14 x 12 in.
2262.77 €
Pack of 250
Please to purchase this item. Need a web account? Register with us today!
30020006
View Documents Promotion Details
Keystone Cleanroom Products
6STK0615SS
15 x 6 in.
1798.48 €
Pack of 250
Please to purchase this item. Need a web account? Register with us today!
30020158
View Documents Promotion Details
Keystone Cleanroom Products
6STK0815SS
15 x 8 in.
2249.23 €
Pack of 250
Please to purchase this item. Need a web account? Register with us today!
30020039
View Documents Promotion Details
Keystone Cleanroom Products
6STK1015SS
15 x 10 in.
2240.23 €
Pack of 250
Please to purchase this item. Need a web account? Register with us today!
30020277
View Documents Promotion Details
Keystone Cleanroom Products
6STK0416SS
16 x 4 in.
1879.62 €
Pack of 250
Please to purchase this item. Need a web account? Register with us today!
30020160
View Documents Promotion Details
Keystone Cleanroom Products
6STK0616SS
16 x 6 in.
1884.13 €
Pack of 250
Please to purchase this item. Need a web account? Register with us today!
30020083
View Documents Promotion Details
Keystone Cleanroom Products
6STK1216SS
16 x 12 in.
2276.29 €
Pack of 250
Please to purchase this item. Need a web account? Register with us today!
30019997
View Documents Promotion Details
Keystone Cleanroom Products
6STK1018SS
18 x 10 in.
2276.29 €
Pack of 250
Please to purchase this item. Need a web account? Register with us today!
30020068
View Documents Promotion Details
Keystone Cleanroom Products
6STK1218SS
18 x 12 in.
2289.81 €
Pack of 250
Please to purchase this item. Need a web account? Register with us today!
30020141
View Documents Promotion Details
Keystone Cleanroom Products
6STK0820SS
20 x 8 in.
2307.85 €
Pack of 250
Please to purchase this item. Need a web account? Register with us today!
30020371
View Documents Promotion Details
Keystone Cleanroom Products
6STK1020SS
20 x 10 in.
2276.29 €
Pack of 250
Please to purchase this item. Need a web account? Register with us today!
30020177
View Documents Promotion Details
Keystone Cleanroom Products
6STK1220SS
20 x 12 in.
2303.33 €
Pack of 250
Please to purchase this item. Need a web account? Register with us today!
30020129
View Documents Promotion Details
Keystone Cleanroom Products
6STK1022SS
22 x 10 in.
2307.85 €
Pack of 250
Please to purchase this item. Need a web account? Register with us today!
30020014
View Documents Promotion Details
Keystone Cleanroom Products
6STK1222SS
22 x 12 in.
2303.33 €
Pack of 250
Please to purchase this item. Need a web account? Register with us today!
30020005
View Documents Promotion Details
Keystone Cleanroom Products
6STK0426SS
26 x 4 in.
1978.79 €
Pack of 250
Please to purchase this item. Need a web account? Register with us today!
30020103
View Documents Promotion Details
Keystone Cleanroom Products
6STK0428SS
28 x 4 in.
2032.88 €
Pack of 250
Please to purchase this item. Need a web account? Register with us today!
30020052
View Documents Promotion Details
Keystone Cleanroom Products
6STK1228SS
28 x 12 in.
2461.10 €
Pack of 250
Please to purchase this item. Need a web account? Register with us today!
30020309
View Documents Promotion Details
Keystone Cleanroom Products
6STK1024SS
24 x 10 in.
2393.48 €
Pack of 250
Please to purchase this item. Need a web account? Register with us today!
30020349
View Documents Promotion Details
Keystone Cleanroom Products
6STK1224SS
24 x 12 in.
2425.04 €
Pack of 250
Please to purchase this item. Need a web account? Register with us today!
30020296
View Documents Promotion Details
Keystone Cleanroom Products
6STK0430SS
30 x 4 in.
2159.10 €
Pack of 250
Please to purchase this item. Need a web account? Register with us today!
30020046
View Documents Promotion Details
Keystone Cleanroom Products
6STK1030SS
30 x 10 in.
2902.83 €
Pack of 250
Please to purchase this item. Need a web account? Register with us today!
30020374
View Documents Promotion Details
Keystone Cleanroom Products
6STK1230SS
30 x 12 in.
2488.13 €
Pack of 250
Please to purchase this item. Need a web account? Register with us today!
30020071
View Documents Promotion Details
Keystone Cleanroom Products
6STK1232SS
32 x 12 in.
2519.69 €
Pack of 250
Please to purchase this item. Need a web account? Register with us today!
30020251
View Documents Promotion Details
Keystone Cleanroom Products
6STK1234SS
34 x 12 in.
2546.73 €
Pack of 250
Please to purchase this item. Need a web account? Register with us today!
30020286
View Documents Promotion Details
Keystone Cleanroom Products
6STK1236SS
36 x 12 in.
2636.88 €
Pack of 250
Please to purchase this item. Need a web account? Register with us today!
30020332
View Documents Promotion Details
Keystone Cleanroom Products
6STK0638SS
38 x 6 in.
2875.79 €
Pack of 250
Please to purchase this item. Need a web account? Register with us today!
30020000
View Documents Promotion Details
Keystone Cleanroom Products
6STK1240SS
40 x 12 in.
2699.98 €
Pack of 250
Please to purchase this item. Need a web account? Register with us today!
30020194
View Documents Promotion Details
Keystone Cleanroom Products
6STK1242SS
42 x 12 in.
2722.52 €
Pack of 250
Please to purchase this item. Need a web account? Register with us today!
30020378
View Documents Promotion Details
Keystone Cleanroom Products
6STK1246SS
46 x 12 in.
2785.63 €
Pack of 250
Please to purchase this item. Need a web account? Register with us today!
30020306
View Documents Promotion Details
Keystone Cleanroom Products
6STK1048SS
48 x 10 in.
3353.58 €
Pack of 250
Please to purchase this item. Need a web account? Register with us today!
30020252
View Documents Promotion Details
Keystone Cleanroom Products
6STK1248SS
48 x 12 in.
2848.73 €
Pack of 250
Please to purchase this item. Need a web account? Register with us today!
30020266
View Documents Promotion Details
Keystone Cleanroom Products
6STK1056SS
56 x 10 in.
3421.19 €
Pack of 250
Please to purchase this item. Need a web account? Register with us today!
30020338
View Documents Promotion Details
Keystone Cleanroom Products
6STK1062SS
62 x 10 in.
3412.17 €
Pack of 250
Please to purchase this item. Need a web account? Register with us today!
Description
Description
  • Designed for steam sterilization
  • Tyvek™ on both sides for maximum breathability during sterilization
  • Integrated steam indicator
  • Recommended for sterilization of pharmaceutical equipment
  • Manufactured and packaged in controlled cleanroom environment
  • Eliminates need to cut material and the use of autoclave tape
  • Self-sealing closure is fast and consistent for equipment preparation area
  • Creates secondary level of protection during aseptic presentation of equipment
  • Resists tearing and punctures
  • Superior microbial barrier
  • Certificates of conformance sent with shipment
Specifications
Specifications
Autoclave Bag
Certificate of Conformance (COC)
White
Steam Sterilization
Steam Sterilization Indicator
Non-sterile
Up to 250°F
5.3 mil
Autoclavable
Self-seal
Single-use
Pharmaceutical Equipment and Components
Tyvek
0.13 mm
Up to 125°C
Videos
SDS
Documents
Documents
Product Certifications