Learn More
Robu™ Series 15 Filter Discs
Robu™ Series 15 Filter Discs feature a unique porous structure, which make them flexible and incredibly versatile for a variety of applications.
Brand: Robu™ 15303
This item is not returnable.
View return policy
Beschreibung
- Surfaces untreated
- Ground cylindrical
- The faces are press molded and may show thickness deviations
Spezifikation
Untreated surface | |
16 to 40 μm | |
3.5 ± 0.8 mm | |
30 mm |
Glass | |
Cylindrical | |
Filter Disc |