Learn More
Memmert™ Stainless Steel Bottom Drip Tray for UF75, 15 mm
Non-perforated bottom drip trays constructed of stainless steel.
Brand: Memmert™ UF75B04358
Description
- Accessory
- May affect temperature distribution
- Constructed of stainless steel
- Rim height: 15 mm
Specifications
UF75 Oven | |
Bottom Drip Tray | |
Stainless Steel | |
Rim: 15 mm |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.