Learn More
Welch™ Stainless Steel KF Inner Centering with Buna N O-ring
9.05 € - 10.05 €
Product Code | Brand | Diameter (Metric) Outside | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) Outside | Price | Quantity & Availability | |||||
10177831
|
Welch™
701061 |
16 mm |
9.05 €
Each |
Estimated Shipment: 09-12-2024
Log in to see stock. |
|||||
10137881
|
Welch™
701063 |
25 mm |
10.05 €
Each |
Estimated Shipment: 03-01-2025
Log in to see stock. |
|||||
Specifications
Stainless Steel | |
1 Each | |
KF Inner Centering |