missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Agencinox™ Stainless Steel Sterilization Boxes
Use these boxes for a variety of general laboratory applications. Agencinox™ Stainless Steel Sterilization Boxes keep instruments safe and secure during sterilization process.
Brand: Agencinox™ 5046
Additional Details : Weight : 0.80000kg
Description
- Stainless steel instrument boxes
- Available in a variety of sizes
Specifications
Stainless Steel | |
Sterilization Box | |
150 mm |
Rectangular | |
250 mm | |
60 mm |