Learn More
Agencinox™ Stainless Steel Sterilization Boxes
Use these boxes for a variety of general laboratory applications. Agencinox™ Stainless Steel Sterilization Boxes keep instruments safe and secure during sterilization process.
65.90 € - 273.00 €
Specifications
Material | Stainless Steel |
---|---|
Shape | Rectangular |
Product Type | Sterilization Box |
Product Code | Brand | Height (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Height (Metric) | Price | Quantity & Availability | |||||
11703924
|
Agencinox™
5031 |
40 mm |
65.90 €
Each |
Estimated Shipment: 22-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11783934
|
Agencinox™
5036 |
50 mm |
81.81 €
Each |
Estimated Shipment: 22-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11793934
|
Agencinox™
5046 |
60 mm |
176.78 €
Each |
Estimated Shipment: 22-07-2024
Log in to see stock. |
|||||
11713944
|
Agencinox™
5075 |
100 mm |
273.00 €
Each |
Estimated Shipment: 20-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11703944
|
Agencinox™
5052 |
70 mm |
174.35 €
Each |
Estimated Shipment: 24-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Stainless steel instrument boxes
- Available in a variety of sizes
Specifications
Stainless Steel | |
Rectangular | |
Sterilization Box |