missing translation for 'onlineSavingsMsg'
Learn More

trans-4-(Benzyloxycarbonylamino)cyclohexylamine, 97%, Thermo Scientific Chemicals

23.10 € - 277.38 €

Chemical Identifiers

CAS 149423-77-8
Molecular Formula C14H20N2O2
Molecular Weight (g/mol) 248.33
MDL Number MFCD03839901,MFCD06657668,MFCD07772066
InChI Key JQVBZZUMWRXDSQ-UHFFFAOYSA-N
Synonym benzyl cis-4-aminocyclohexyl carbamate, benzyl trans-4-aminocyclohexyl carbamate, n-cbz-trans-1,4-cyclohexanediamine, cis-1-n-cbz-1,4-cyclohexyldiamine, cis-n-cbz-1,4-cyclohexanediamine, 4-amino-cyclohexyl-carbamic acid benzyl ester, benzyl cis-4-aminocyclohexylcarbamate, benzyl 4-aminocyclohexyl carbamate, benzyl 1r,4r-4-aminocyclohexyl carbamate, 1-n-cbz-trans-1,4-cyclohexyldiamine
PubChem CID 18650082
IUPAC Name benzyl N-(4-aminocyclohexyl)carbamate
SMILES NC1CCC(CC1)NC(=O)OCC1=CC=CC=C1
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15447446
SDS View Documents
Thermo Scientific Alfa Aesar
H62444.MD
250 mg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15427446
SDS View Documents
Thermo Scientific Alfa Aesar
H62444.03
1 g
71.86 €
1g
Please to purchase this item. Need a web account? Register with us today!
15437446
SDS View Documents
Thermo Scientific Alfa Aesar
H62444.06
5 g
277.38 €
5g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

trans-4-(Benzyloxycarbonylamino)cyclohexylamine is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
trans-4-(Benzyloxycarbonylamino)cyclohexylamine is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.
Specifications
Show More
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
trans-4-(Benzyloxycarbonylamino)cyclohexylamine, 97%, Thermo Scientific Chemicals

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.