missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ UltraVision LP Detection System

Epredia™ Lab Vision UltraVision LP Detection System: HRP Polymer is a sensitive detection system based on biotin-free, polymer technology.

Brand:  Epredia™ TL-060-HL

 View more versions of this product

Product Code. 10627585

This item is not returnable. View return policy

Description

Description

UltraVision LP is the latest technology in polymeric labeling. Polymer detection methods have been shown to provide increased sensitivity and detection simplicity. This second-generation polymer system is composed of smaller polymer subunits that minimize conflicts in binding the target protein. Decreased binding conflicts result in more consistent staining and better signal amplification.1 Ultimately, this gives the user higher sensitivity and antibody efficiency.2 With UltraVision LP, you use less antibody and obtain better signal-to-noise ratios. UltraVision LP is also biotin-free, which eliminates background staining found with traditional biotin-based detection methods.
  • Specificity: anti-Mouse IgG (H+L), anti-Rabbit IgG (H+L)

  • Enzyme: Peroxidase

  • Chromogen/Substrate: None provided

Includes:

TL-060-HL: 60mL Ultra V Block (TA-060-UB), 60mL Primary Antibody Enhancer (TL-060-PB), 60mL HRP Polymer (TL-060-PH)

TL-125-HL: 125mL Ultra V Block (TA-125-UB), 125mL Primary Antibody Enhancer (TL-125-PB), 125mL HRP Polymer (TL-125-PH)

Note:

UltraVision detection system detects a specific mouse IgG or rabbit IgG antibody bound to an antigen in tissue sections. The specific antibody is located by a universal secondary antibody formulation conjugated to an enzyme-labeled polymer that recognizes mouse and rabbit immunoglobulins. The polymer complex is then visualized with an appropriate substrate/chromogen.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

300 to 600 slides
60mL Ultra V Block (TA-060-UB), 60mL Primary Antibody Enhancer (TL-060-PB), 60mL HRP Polymer (TL-060-PH)
anti-Mouse IgG (H+L), anti-Rabbit IgG (H+L)
Peroxidase
None provided
60 mL
None provided
Store at 2° to 8°C. Each component is stable for 18 months.
Lab Vision™, UltraVision™
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

One moment while we fetch your results.
Certificados
Ofertas especiales

Ofertas especiales

Corrección del contenido de un producto

Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.

Título del producto
Epredia™ UltraVision LP Detection System > 60mL

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.

Gracias por ayudarnos a mejorar nuestra web. Su comentario ha sido enviado