Learn More
Idex ZDV Low-Pressure Delrin Acetal Unions
Make solid connections in low-pressure applications
Brand:  Idex 02013-94
 View more versions of this product
Product Code. 12967719
  • 6.10 € / Each
Estimated Shipment: 21-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Unions do not include fittings—provide your own to match your tubing size
  • Manufactured from inert polymers
  • True ZDV union
  • Acetal body
Specifications
Specifications
Union
60°C
Acetal
White