Learn More
Idex ZDV Low-Pressure Delrin Acetal Unions
Make solid connections in low-pressure applications
6.10 €
Specifications
Type Union
Temperature (Metric) Max 60°C
Material Acetal
View More Specs
Products 1
Product Code Brand Color Price Quantity & Availability  
Product Code Brand Color Price Quantity & Availability  
12967719
View Documents
Idex
02013-94
White
6.10 €
Each
Estimated Shipment: 07-01-2025
to see stock.
Add to basket
Description
Description
  • Unions do not include fittings—provide your own to match your tubing size
  • Manufactured from inert polymers
  • True ZDV union
  • Acetal body
Specifications
Specifications
Union
60°C
Acetal