Learn More
Memmert™ USB User-ID Stick (with User-ID Licence) for UN260plus
USB User-ID Stick prevents undesired manipulation by unauthorized third parties.
Brand: Memmert™ UN260plusB33170
Description
- Oven-linked authorization licence (User-ID-program) on Memory-stick
- Prevents undesired manipulation by unauthorised third parties
- When reordering please specify serial number (only for units with TwinDISPLAY)
Specifications
UN260plus Oven | |
USB Stick | |
User ID Licence |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.