Learn More
Advantage-Lab™ UV Lamp, 2 x 15 Watt
Useful for detecting fluorescent particles and minerals, curing adhesives and paints, and testing coatings. Advantage-Lab™ UV Lamp 2 x 15 Watt is a high-intensity, single-wave unit designed with an integrated handle and a powder-coated, metal housing.
Brand: Advantage-Lab™ AL10-30-365
![Encompass](/content/dam/fishersci/glyphs/encompass.png)
This item is not returnable.
View return policy
Description
- Useful in many fields including the following:
- Molecular biology.
- Microbiology.
- Botany.
- Chemistry.
- Mineralogy.
- Glass production.
- High intensity.
- Single-wave unit.
- Powder-coated metal housing.
- Integrated handle.
- Adjustable stand (option).
Specifications
220/240 V (optional 110/120 V) | |
Longwave UV-A | |
1400 μW/cm2 |
2 x 15 W | |
485 x 85 x 80 mm |