Learn More
Advantage-Lab™ UV Lamp, 2 x 15 Watt
Useful for detecting fluorescent particles and minerals, curing adhesives and paints, and testing coatings. Advantage-Lab™ UV Lamp 2 x 15 Watt is a high-intensity, single-wave unit designed with an integrated handle and a powder-coated, metal housing.
901.00 € - 1030.00 €
Specifications
Voltage 220/240 V (optional 110/120 V)
Dimensions (L x W x H) 485 x 85 x 80 mm
View More Specs
Products 5
Product Code Brand Lighting Intensity Wavelength Price Quantity & Availability  
Product Code Brand Lighting Intensity Wavelength Price Quantity & Availability  
15608905
Encompass
View Documents
Advantage-Lab™
AL10-30-254
1400 μW/cm2 Shortwave UV-C
998.00 €
Each
View alternative products
Save Up to 
»
Estimated Shipment: 16-01-2025
to see stock.
Add to basket
15618905
Encompass
View Documents
Advantage-Lab™
AL10-30-312
1300 μW/cm2 Midwave UV-B
1030.00 €
Each
View alternative products
Save Up to 
»
Estimated Shipment: 16-01-2025
to see stock.
Add to basket
15628905
Encompass
View Documents
Advantage-Lab™
AL10-30-365
1400 μW/cm2 Longwave UV-A
901.00 €
Each
View alternative products
Save Up to 
»
Estimated Shipment: 16-01-2025
to see stock.
Add to basket
15658895
Encompass
View Documents
Advantage-Lab™
AL10-15-566
1200/1100 μW/cm2 Short/Midwave
960.00 €
Each
View alternative products
Save Up to 
»
Estimated Shipment: 16-01-2025
to see stock.
Add to basket
15668895
Encompass
View Documents
Advantage-Lab™
AL10-15-619
1200/1300 μW/cm2 Short/Longwave
932.00 €
Each
View alternative products
Save Up to 
»
Estimated Shipment: 16-01-2025
to see stock.
Add to basket
Description
Description
  • Useful in many fields including the following:
    • Molecular biology.
    • Microbiology.
    • Botany.
    • Chemistry.
    • Mineralogy.
    • Glass production.
  • High intensity.
  • Single-wave unit.
  • Powder-coated metal housing.
  • Integrated handle.
  • Adjustable stand (option).
Specifications
Specifications
220/240 V (optional 110/120 V)
485 x 85 x 80 mm
Videos
SDS
Documents
Documents
Product Certifications