Learn More
Advantage-Lab™ UV Lamp, 2 x 15 Watt
Useful for detecting fluorescent particles and minerals, curing adhesives and paints, and testing coatings. Advantage-Lab™ UV Lamp 2 x 15 Watt is a high-intensity, single-wave unit designed with an integrated handle and a powder-coated, metal housing.
Brand:  Advantage-Lab™ AL10-15-619
Encompass
 View more versions of this product
Product Code. 15668895
  • 932.00 € / Each
Estimated Shipment: 17-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy
Description
Description
  • Useful in many fields including the following:
    • Molecular biology.
    • Microbiology.
    • Botany.
    • Chemistry.
    • Mineralogy.
    • Glass production.
  • High intensity.
  • Single-wave unit.
  • Powder-coated metal housing.
  • Integrated handle.
  • Adjustable stand (option).
Specifications
Specifications
220/240 V (optional 110/120 V)
Short/Longwave
1200/1300 μW/cm2
1 x 15 W
485 x 85 x 80 mm