Learn More
Advantage-Lab™ UV Lamp, 2 x 8 Watt
Useful for detecting fluorescent particles and minerals, curing adhesives and paints, and testing coatings. Advantage-Lab™ UV Lamp 2 x 8 Watt is a high-intensity, single-wave unit designed with an integrated handle and a powder-coated, metal housing.
Brand:  Advantage-Lab™ AL10-16-254
Encompass
 View more versions of this product
Product Code. 15678895
  • 709.00 € / Each
Estimated Shipment: 18-02-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy
Description
Description
  • Useful in many fields:
    • Molecular biology.
    • Microbiology.
    • Botany.
    • Chemistry.
    • Mineralogy.
    • Glass production.
  • High intensity.
  • Single-wave unit.
  • Powder-coated metal housing.
  • Integrated handle.
  • Adjustable stand (option).
Specifications
Specifications
220/240 V (optional 110/120 V)
Shortwave UV-C
1300 μW/cm2
2 x 8 W
335 x 70 x 65 mm