Learn More
Advantage-Lab™ UV Lamp, 2 x 8 Watt
Useful for detecting fluorescent particles and minerals, curing adhesives and paints, and testing coatings. Advantage-Lab™ UV Lamp 2 x 8 Watt is a high-intensity, single-wave unit designed with an integrated handle and a powder-coated, metal housing.
695.00 € - 730.00 €
Specifications
Voltage 220/240 V (optional 110/120 V)
Dimensions (L x W x H) 335 x 70 x 65 mm
View More Specs
Products 5
Product Code Brand Lighting Intensity Wavelength Price Quantity & Availability  
Product Code Brand Lighting Intensity Wavelength Price Quantity & Availability  
15698895
Encompass
missing translation for 'viewDocuments'
Advantage-Lab™
AL10-16-365
1900 μW/cm2 Longwave UV-A
695.00 €
Each
View alternative products
missing translation for 'toSaveUpto' 
»
Estimated Shipment: 18-02-2025
to see stock.
Add to basket
15678885
Encompass
missing translation for 'viewDocuments'
Advantage-Lab™
AL10-08-566
680/650 μW/cm2 Short/Midwave
713.00 €
Each
View alternative products
missing translation for 'toSaveUpto' 
»
Estimated Shipment: 18-02-2025
to see stock.
Add to basket
15678895
Encompass
missing translation for 'viewDocuments'
Advantage-Lab™
AL10-16-254
1300 μW/cm2 Shortwave UV-C
709.00 €
Each
View alternative products
missing translation for 'toSaveUpto' 
»
Estimated Shipment: 18-02-2025
to see stock.
Add to basket
15688885
Encompass
missing translation for 'viewDocuments'
Advantage-Lab™
AL10-08-619
680/950 μW/cm2 Short/Longwave
704.00 €
Each
View alternative products
missing translation for 'toSaveUpto' 
»
Estimated Shipment: 18-02-2025
to see stock.
Add to basket
15688895
Encompass
missing translation for 'viewDocuments'
Advantage-Lab™
AL10-16-312
1250 μW/cm2 Midwave UV-B
730.00 €
Each
View alternative products
missing translation for 'toSaveUpto' 
»
Estimated Shipment: 18-02-2025
to see stock.
Add to basket
Description
Description
  • Useful in many fields:
    • Molecular biology.
    • Microbiology.
    • Botany.
    • Chemistry.
    • Mineralogy.
    • Glass production.
  • High intensity.
  • Single-wave unit.
  • Powder-coated metal housing.
  • Integrated handle.
  • Adjustable stand (option).
Specifications
Specifications
220/240 V (optional 110/120 V)
335 x 70 x 65 mm