Learn More
Advantage-Lab™ UV Lamp, 2 x 8 Watt
Useful for detecting fluorescent particles and minerals, curing adhesives and paints, and testing coatings. Advantage-Lab™ UV Lamp 2 x 8 Watt is a high-intensity, single-wave unit designed with an integrated handle and a powder-coated, metal housing.
695.00 € - 730.00 €
Specifications
Voltage | 220/240 V (optional 110/120 V) |
---|---|
Dimensions (L x W x H) | 335 x 70 x 65 mm |
Product Code | Brand | Lighting Intensity | Wavelength | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Lighting Intensity | Wavelength | Price | Quantity & Availability | ||||
15698895
|
Advantage-Lab™
AL10-16-365 |
1900 μW/cm2 | Longwave UV-A |
695.00 €
Each |
Estimated Shipment: 18-02-2025
Log in to see stock. |
||||
15678885
|
Advantage-Lab™
AL10-08-566 |
680/650 μW/cm2 | Short/Midwave |
713.00 €
Each |
Estimated Shipment: 18-02-2025
Log in to see stock. |
||||
15678895
|
Advantage-Lab™
AL10-16-254 |
1300 μW/cm2 | Shortwave UV-C |
709.00 €
Each |
Estimated Shipment: 18-02-2025
Log in to see stock. |
||||
15688885
|
Advantage-Lab™
AL10-08-619 |
680/950 μW/cm2 | Short/Longwave |
704.00 €
Each |
Estimated Shipment: 18-02-2025
Log in to see stock. |
||||
15688895
|
Advantage-Lab™
AL10-16-312 |
1250 μW/cm2 | Midwave UV-B |
730.00 €
Each |
Estimated Shipment: 18-02-2025
Log in to see stock. |
||||
Description
- Useful in many fields:
- Molecular biology.
- Microbiology.
- Botany.
- Chemistry.
- Mineralogy.
- Glass production.
- High intensity.
- Single-wave unit.
- Powder-coated metal housing.
- Integrated handle.
- Adjustable stand (option).
Specifications
220/240 V (optional 110/120 V) | |
335 x 70 x 65 mm |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.