Learn More
Masterflex™ Viton™ L/S™ 2-Stop Pump Tubing
Good for food and lab applications
Brand:  Masterflex™ 96428-16
 View more versions of this product
Product Code. 11750159
  • 250.00 € / Pack of 8
Estimated Shipment: 02-06-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy
Description
Description
  • Genuine Masterflex™ tubing undergoes robust testing to provide optimal performance / pump life hours with peristaltic pump equipment
  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimize downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include packs, cases, and spools for customer-friendly ordering and delivery
  • Food or laboratory applications where FDA compliance is required, acid and solvent transfer, high-temperature work
  • Good chemical resistance, opaque black
  • For use with Masterflex™ multichannel pump heads
Specifications
Specifications
FDA-Compliant
Viton
0.38 m
1 / Pk.
460 mL/min
660 mm Hg
Stopped Pump Tubing
3.1 mm
1.25 ft.
0°C
16
1/8 in.
150°C
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Masterflex™ Viton™ L/S™ 2-Stop Pump Tubing > 16