Learn More
Emma Safety Footwear Zion Safety Shoes
Chemical resistant with highly visible 3M™ reflective strip. The impenetrable sole protects feet from sharp and metal objects while the steel toe cap and reinforced polyurethane toe cover offer protection from falling objects. Microfiber components stand up to cement and remains flexible.
Brand: Emma Safety Footwear MM30084840
This item is not returnable.
View return policy
Description
- S3-Safety shoe with sole plate for wet work
- Microfiber construction
- GripForce™ Duo Heat Resistant Outsole
- 3M™ Reflective Striping
- Hydro-Tec™ Sanitized Silver provides drier, fresher feet
Specifications
EN ISO 20345:2011 | |
Zion | |
Microfiber |
Black | |
Hydro-Tec™ Sanitized Silver | |
40 |