Learn More
Memmert™ Entry Port 100 mm for UN260plus
Optional entry port for Memmert Ovens.
Brand:  Memmert™ UN260plusF9
Product Code. 17520036
  • 590.00 € / Each
Estimated Shipment: 14-05-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy
Description
Description
  • Entry port with 100 mm diameter
  • Flap closure
  • Custom position TBC in the back
Specifications
Specifications
UN260plus Oven
Entry Port
100 mm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Memmert™ Entry Port 100 mm for UN260plus > For Use With: UN260plus