Learn More
Memmert™ Flexible Pt100 Temperature Sensor for UF750
Flexible temperature sensor for positioning in chamber or in load.
Brand:  Memmert™ UF750H8
Product Code. 17589565
  • 226.00 € / Each
Estimated Shipment: 23-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Positioned flexibly in chamber or load
  • For local temperature measurement (up to 1 additional sensor is possible)
  • The measured temperature will be indicated on the display, recorded in the integral data store, and can be documented via the AtmoCONTROL software
  • For Memmert Ovens
Specifications
Specifications
UF750 Oven
Temperature Sensor
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Memmert™ Flexible Pt100 Temperature Sensor for UF750 > For Use With: UF750