missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Glypican 1 Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems MAB45192-SP
This item is not returnable.
View return policy
Description
Glypican 1 Monoclonal antibody specifically detects Glypican 1 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry
Specifications
Glypican 1 | |
Monoclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
FLJ38078, glypican, glypican 1, glypican proteoglycan 1, glypican-1, GPC1 | |
Mouse myeloma cell line NS0derived human Glypican 1, Asp24-Ser530, Accession # P35052 | |
25 μg | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. | |
IgG1 |
Flow Cytometry, Immunocytochemistry | |
1019718 | |
Flow Cytometry 0.25 ug/10^6 cells, Immunocytochemistry 8-25 ug/mL | |
P35052 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
2817 | |
Human | |
Purified |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-5 of 12
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Human Glypican 1 Antibody, R&D Systems™ > 25 μg; Unconjugated
Spot an opportunity for improvement?Share a Content Correction